siRNA Details
| Gene Symbol: Evi2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033711.1 | NP_001028883.1 | 2 | 2184 | ||
| NM_010161.3 | NP_034291.1 | 2 | 1371 | |||
| GenBank mRNA | AK005353.1 | - | 2 | 1348 | ||
| AK044098.1 | - | 2 | 1247 | |||
| AK046407.1 | - | 2 | 1322 | |||
| AK160302.1 | - | 2 | 1288 | |||
| AK162958.1 | - | 1247 | ||||
| AK170527.1 | - | 2 | 1343 | |||
| AK171594.1 | - | 2 | 1289 | |||
| BC038124.1 | - | 2 | 1328 | 1396641 | ||

Evi2a
Genomic Map