siRNA Details
| Gene Symbol: AKAP17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005088.2 | NP_005079.2 | 2 | 732 | ||
| NR_027383.1 | 2 | 732 | ||||
| GenBank mRNA | AK308367.1 | - | 2 | 725 | ||
| BC016935.1 | - | 2 | 732 | |||
| BC028151.1 | - | 2 | 700 | 5217247 | ||
| BC110496.1 | - | 2 | 730 | 40007662 | ||
| BC110497.1 | - | 2 | 730 | 40007663 | ||
| L03426.1 | - | 2 | 702 | |||
| M99578.1 | - | 2 | 705 | |||

AKAP17A
Genomic Map