siRNA Details
| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252192.1 | NP_001239121.1 | 3 | 631 | ||
| NM_010164.2 | NP_034294.2 | 4 | 759 | |||
| GenBank mRNA | AK042412.1 | - | 3 | 554 | ||
| BC060260.1 | - | 4 | 759 | 6848408 | ||
| BC066860.1 | - | 4 | 416 | 30089692 | ||
| U61110.1 | - | 3 | 304 | |||

Eya1
Genomic Map