siRNA Details
| Gene Symbol: Fcgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010186.5 | NP_034316.1 | 6 | 2467 | ||
| GenBank mRNA | AK033874.1 | - | 6 | 2467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcgr1| Gene Symbol: Fcgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010186.5 | NP_034316.1 | 6 | 2467 | ||
| GenBank mRNA | AK033874.1 | - | 6 | 2467 | ||