siRNA Details
| Gene Symbol: Fcgr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK169887.1 | - | 5 | 1058 | ||
| AK171289.1 | - | 5 | 1062 | |||
| M14215.1 | - | 5 | 1095 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcgr3| Gene Symbol: Fcgr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK169887.1 | - | 5 | 1058 | ||
| AK171289.1 | - | 5 | 1062 | |||
| M14215.1 | - | 5 | 1095 | |||