siRNA Details
| Gene Symbol: ZMYM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171162.1 | NP_001164633.1 | 4 | 1513 | ||
| NM_001171163.1 | NP_001164634.1 | 4 | 952 | |||
| NM_005096.3 | NP_005087.1 | 4 | 952 | |||
| NM_201599.2 | NP_963893.1 | 4 | 943 | |||
| XM_005262309.3 | XP_005262366.1 | 4 | 1386 | |||
| XM_005262310.2 | XP_005262367.1 | 4 | 899 | |||
| XM_011531062.2 | XP_011529364.1 | 4 | 852 | |||
| GenBank mRNA | AB002383.1 | - | 4 | 851 | ||
| AK290762.1 | - | 4 | 952 | |||
| BC013009.2 | - | 4 | 876 | |||
| BC069057.1 | - | 4 | 856 | 5499547 | ||
| BT007095.1 | - | 3 | 764 | |||
| X95808.1 | - | 5 | 1489 | |||

ZMYM3
Genomic Map