siRNA Details
| Gene Symbol: Fem1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010192.4 | NP_034322.3 | 1 | 3874 | ||
| GenBank mRNA | AI021455.1 | - | 1 | 104 | ||
| AK080978.1 | - | 1 | 3847 | |||
| CO426831.1 | - | 1 | 52 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fem1a| Gene Symbol: Fem1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010192.4 | NP_034322.3 | 1 | 3874 | ||
| GenBank mRNA | AI021455.1 | - | 1 | 104 | ||
| AK080978.1 | - | 1 | 3847 | |||
| CO426831.1 | - | 1 | 52 | |||