siRNA Details
| Gene Symbol: Fem1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010193.4 | NP_034323.1 | 1 | 429 | ||
| GenBank mRNA | AB022863.1 | - | 1 | 339 | ||
| AF064448.1 | - | 1 | 160 | |||
| AF178633.1 | - | 1 | 86 | |||
| AK032338.1 | - | 1 | 406 | |||
| AK132692.1 | - | 1 | 422 | |||
| AK149329.1 | - | 1 | 392 | |||
| AK154060.1 | - | 1 | 717 | |||
| BC068236.1 | - | 1 | 383 | 30544277 | ||

Fem1b
Genomic Map