siRNA Details
| Gene Symbol: Fem1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010193.4 | NP_034323.1 | 2 | 3558 | ||
| GenBank mRNA | AK122272.1 | - | 1 | 2754 | ||
| AK145371.1 | - | 1 | 2919 | |||
| AK160393.1 | - | 1 | 1494 | |||
| BC068236.1 | - | 2 | 3513 | 30544277 | ||

Fem1b
Genomic Map