siRNA Details
| Gene Symbol: EXOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318956.1 | NP_001305885.1 | 2 | 323 | ||
| NM_001318957.1 | NP_001305886.1 | 2 | 323 | |||
| NM_001318959.1 | NP_001305888.1 | 3 | 410 | |||
| NM_005107.3 | NP_005098.2 | 2 | 323 | |||
| XM_005265635.3 | XP_005265692.1 | 2 | 272 | |||
| XM_006713442.3 | XP_006713505.1 | 3 | 555 | |||
| XM_017007584.1 | XP_016863073.1 | 2 | 250 | |||
| XM_017007585.1 | XP_016863074.1 | 2 | 250 | |||
| XM_017007586.1 | XP_016863075.1 | 2 | 250 | |||
| XM_017007587.1 | XP_016863076.1 | 2 | 240 | |||
| XM_017007588.1 | XP_016863077.1 | 3 | 555 | |||
| XM_017007589.1 | XP_016863078.1 | 3 | 555 | |||
| XM_017007590.1 | XP_016863079.1 | 2 | 101 | |||
| XM_017007591.1 | XP_016863080.1 | 2 | 109 | |||
| GenBank mRNA | AB020523.1 | - | 2 | 254 | ||
| AK023235.1 | - | 2 | 230 | |||
| AK315814.1 | - | 3 | 185 | |||
| BC104212.1 | - | 2 | 252 | 40031649 | ||

EXOG
Genomic Map