siRNA Details
| Gene Symbol: Fmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285458.1 | NP_001272387.1 | 4 | 2914 | ||
| NM_001285459.1 | NP_001272388.1 | 4 | 2538 | |||
| NM_010230.2 | NP_034360.2 | 6 | 3208 | |||
| GenBank mRNA | BC138036.1 | - | 3 | 2820 | 8861055 | |
| BC171945.1 | - | 3 | 2814 | 9054231 | ||
| X53599.1 | - | 6 | 3168 | |||
| X62379.1 | - | 4 | 2544 | |||

Fmn1
Genomic Map