siRNA Details
| Gene Symbol: Fth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010239.2 | NP_034369.1 | 4 | 639 | ||
| GenBank mRNA | AK027998.1 | - | 4 | 582 | ||
| AK088935.1 | - | 3 | 1787 | |||
| AK139622.1 | - | 4 | 582 | |||
| AK147082.1 | - | 4 | 585 | |||
| AK150262.1 | - | 4 | 582 | |||
| AK150508.1 | - | 4 | 582 | |||
| AK150628.1 | - | 4 | 582 | |||
| AK150679.1 | - | 4 | 585 | |||
| AK150693.1 | - | 4 | 582 | |||
| AK151192.1 | - | 4 | 582 | |||
| AK151241.1 | - | 4 | 585 | |||
| AK151399.1 | - | 4 | 568 | |||
| AK151609.1 | - | 4 | 582 | |||
| AK151675.1 | - | 4 | 582 | |||
| AK152071.1 | - | 4 | 582 | |||
| AK152542.1 | - | 4 | 585 | |||
| AK152702.1 | - | 5 | 1210 | |||
| AK153017.1 | - | 4 | 582 | |||
| AK153195.1 | - | 4 | 585 | |||
| AK153199.1 | - | 4 | 585 | |||
| AK159243.1 | - | 4 | 582 | |||
| AK168601.1 | - | 4 | 582 | |||
| AK169004.1 | - | 4 | 582 | |||
| BC011096.1 | - | 4 | 565 | |||
| BC012314.1 | - | 4 | 560 | 3488821 | ||
| J03941.1 | - | 4 | 535 | |||
| X12812.1 | - | 4 | 481 | |||

Fth1
Genomic Map