siRNA Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146459.1 | - | 10 | 1313 | ||
| BC008130.1 | - | 11 | 1318 | 3498984 | ||
| X71978.1 | - | 10 | 1337 | |||
| Z67963.1 | - | 10 | 1337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aktip| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146459.1 | - | 10 | 1313 | ||
| BC008130.1 | - | 11 | 1318 | 3498984 | ||
| X71978.1 | - | 10 | 1337 | |||
| Z67963.1 | - | 10 | 1337 | |||