siRNA Details
| Gene Symbol: Fut9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010243.3 | NP_034373.1 | 3 | 3261 | ||
| GenBank mRNA | AK047650.1 | - | 3 | 3060 | ||
| AK163871.1 | - | 3 | 3061 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fut9| Gene Symbol: Fut9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010243.3 | NP_034373.1 | 3 | 3261 | ||
| GenBank mRNA | AK047650.1 | - | 3 | 3060 | ||
| AK163871.1 | - | 3 | 3061 | |||