siRNA Details
| Gene Symbol: Gabra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010250.5 | NP_034380.1 | 10 | 3053 | ||
| GenBank mRNA | AK034095.1 | - | 10 | 3054 | ||
| AK039496.1 | - | 10 | 2337 | |||
| AK141596.1 | - | 10 | 2913 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra1| Gene Symbol: Gabra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010250.5 | NP_034380.1 | 10 | 3053 | ||
| GenBank mRNA | AK034095.1 | - | 10 | 3054 | ||
| AK039496.1 | - | 10 | 2337 | |||
| AK141596.1 | - | 10 | 2913 | |||