siRNA Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010260.1 | NP_034390.1 | 10 | 1783 | ||
| GenBank mRNA | AF077007.1 | - | 10 | 1783 | ||
| AF109168.1 | - | 10 | 1736 | |||
| AJ007970.1 | - | 10 | 1776 | |||
| AK150146.1 | - | 6 | 1085 | |||
| BC011336.1 | - | 10 | 1772 | 3494311 | ||
| BC032882.1 | - | 10 | 1798 | 1230883 | ||
| BC049141.1 | - | 10 | 1760 | |||
| CT010249.1 | - | 9 | 1613 | |||

Gbp2
Genomic Map