siRNA Details
| Gene Symbol: Gdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010267.3 | NP_034397.1 | 3 | 430 | ||
| GenBank mRNA | AK020988.1 | - | 3 | 414 | ||
| AK045460.1 | - | 3 | 430 | |||
| AK049655.1 | - | 3 | 400 | |||
| AK083814.1 | - | 3 | 431 | |||
| AK139057.1 | - | 3 | 273 | |||
| AK149281.1 | - | 3 | 441 | |||
| BC048177.1 | - | 3 | 406 | 6825756 | ||
| BC051135.1 | - | 3 | 395 | 6532839 | ||
| BC060212.1 | - | 3 | 403 | |||
| Y17850.1 | - | 3 | 418 | |||

Gdap1
Genomic Map