siRNA Details
| Gene Symbol: Gdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010267.3 | NP_034397.1 | 6 | 1967 | ||
| GenBank mRNA | AK083814.1 | - | 6 | 1968 | ||
| BC048177.1 | - | 6 | 1943 | 6825756 | ||
| BC051135.1 | - | 6 | 1932 | 6532839 | ||
| BC060212.1 | - | 6 | 1941 | |||
| Y17850.1 | - | 6 | 1956 | |||

Gdap1
Genomic Map