siRNA Details
| Gene Symbol: Gfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267621.1 | NP_001254550.1 | 7 | 1450 | ||
| NM_010278.2 | NP_034408.1 | 6 | 1662 | |||
| GenBank mRNA | AK089065.1 | - | 7 | 1451 | ||
| AK132814.1 | - | 1 | 318 | |||
| BC130219.1 | - | 5 | 847 | |||
| BC152328.1 | - | 5 | 738 | |||
| BU698904.1 | - | 4 | 478 | |||
| U58972.1 | - | 6 | 1637 | |||
| U78312.1 | - | 6 | 1662 | |||

Gfi1
Genomic Map