siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048178.2 | NP_001041643.1 | 4 | 520 | ||
| NM_001286370.1 | NP_001273299.1 | 4 | 221 | |||
| NM_010284.3 | NP_034414.2 | 4 | 520 | |||
| GenBank mRNA | AK053579.1 | - | 4 | 381 | ||
| AK143886.1 | - | 4 | 222 | |||
| AK143925.1 | - | 4 | 318 | |||
| BC024375.1 | - | 4 | 355 | |||
| BC056645.1 | - | 4 | 336 | |||
| BC075720.1 | - | 4 | 368 | 5044060 | ||
| BY754177.1 | - | 4 | 521 | |||
| M31680.1 | - | 4 | 256 | |||
| M33324.1 | - | 4 | 256 | |||

Ghr
Genomic Map