siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286370.1 | NP_001273299.1 | 11 | 3225 | ||
| NM_010284.3 | NP_034414.2 | 11 | 3524 | |||
| GenBank mRNA | AK143925.1 | - | 11 | 3322 | ||
| BC075720.1 | - | 11 | 3372 | 5044060 | ||
| BE625087.1 | - | 1 | 374 | |||

Ghr
Genomic Map