siRNA Details
| Gene Symbol: Gm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010299.3 | NP_034429.1 | 4 | 1045 | ||
| GenBank mRNA | AK149642.1 | - | 4 | 938 | ||
| AK152256.1 | - | 4 | 937 | |||
| BC004651.1 | - | 4 | 913 | 3482848 | ||
| U09816.1 | - | 4 | 922 | |||

Gm2a
Genomic Map