siRNA Details
| Gene Symbol: Gna13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010303.3 | NP_034433.3 | 4 | 2452 | ||
| GenBank mRNA | AK078125.1 | - | 1 | 1381 | ||
| AK149766.1 | - | 4 | 2346 | |||
| BC057665.1 | - | 5 | 2220 | 4918717 | ||
| M63660.1 | - | 4 | 2400 | |||

Gna13
Genomic Map