siRNA Details
Gene Symbol: Gng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010316.3 | NP_034446.1 | 2, 3 | 348 | ||
GenBank mRNA | AK004459.1 | - | 2, 3 | 313 | ||
AK032646.1 | - | 2, 3 | 296 | |||
BC029680.1 | - | 2, 3 | 232 | 5365311 | ||
U38497.1 | - | 1, 2 | 55 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
