siRNA Details
Gene Symbol: Gp1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010326.2 | NP_034456.2 | 2 | 1115 | ||
GenBank mRNA | BC137765.1 | - | 2 | 1095 | 8860784 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gp1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010326.2 | NP_034456.2 | 2 | 1115 | ||
GenBank mRNA | BC137765.1 | - | 2 | 1095 | 8860784 |