siRNA Details
| Gene Symbol: Gpaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010331.2 | NP_034461.1 | 10 | 1485 | ||
| GenBank mRNA | AB002136.1 | - | 10 | 1414 | ||
| AB006970.1 | - | 10 | 1421 | |||
| AK077518.1 | - | 10 | 1485 | |||
| AK164219.1 | - | 10 | 1431 | |||
| AK164978.1 | - | 10 | 1485 | |||
| AK168057.1 | - | 10 | 1688 | |||
| AK177221.1 | - | 2 | 154 | |||
| BC006697.1 | - | 10 | 1424 | 3498573 | ||

Gpaa1
Genomic Map