siRNA Details
Gene Symbol: Gpr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF065145.1 | - | 2 | 389 | ||
AK132272.1 | - | 2 | 490 | |||
BC137982.1 | - | 2 | 386 | 8861001 | ||
BC145080.1 | - | 2 | 386 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF065145.1 | - | 2 | 389 | ||
AK132272.1 | - | 2 | 490 | |||
BC137982.1 | - | 2 | 386 | 8861001 | ||
BC145080.1 | - | 2 | 386 |