siRNA Details
| Gene Symbol: Gsta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077353.2 | NP_001070821.1 | 4 | 319 | ||
| NM_001288617.1 | NP_001275546.1 | 4 | 364 | |||
| NM_010356.4 | NP_034486.2 | 3 | 224 | |||
| GenBank mRNA | AI047326.1 | - | 4 | 297 | ||
| AI255699.1 | - | 3 | 220 | |||
| AK002478.1 | - | 2 | 172 | |||
| AK014076.1 | - | 3 | 197 | |||
| AK149543.1 | - | 3 | 189 | |||
| BC147272.1 | - | 3 | 187 | 9056327 | ||
| BC147273.1 | - | 3 | 187 | 9056339 | ||
| GQ227791.1 | - | 2 | 149 | |||
| M73483.1 | - | 2 | 155 | |||
| X65021.1 | - | 2 | 140 | |||

Gsta3
Genomic Map