siRNA Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010369.3 | NP_034499.3 | 6 | 372 | ||
| GenBank mRNA | AK145998.1 | - | 6 | 373 | ||
| AK157851.1 | - | 6 | 373 | |||
| AK162614.1 | - | 6 | 357 | |||
| BC148192.1 | - | 6 | 636 | 40045496 | ||

Gypa
Genomic Map