siRNA Details
| Gene Symbol: Hba-x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010405.4 | NP_034535.1 | 2 | 300 | ||
| GenBank mRNA | AK077590.1 | - | 2 | 299 | ||
| BC051988.1 | - | 2 | 270 | 5711898 | ||
| BY111754.1 | - | 2 | 301 | |||
| M13125.1 | - | 1 | 108 | |||
| M26898.1 | - | 2 | 258 | |||

Hba-x
Genomic Map