siRNA Details
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010406.2 | NP_034536.1 | 1, 2 | 131 | ||
| GenBank mRNA | AK080166.1 | - | 1, 2 | 130 | ||
| M35525.1 | - | 1, 2 | 86 | |||
| M35526.1 | - | 1, 2 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hc| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010406.2 | NP_034536.1 | 1, 2 | 131 | ||
| GenBank mRNA | AK080166.1 | - | 1, 2 | 130 | ||
| M35525.1 | - | 1, 2 | 86 | |||
| M35526.1 | - | 1, 2 | 86 | |||