siRNA Details
Gene Symbol: Hc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010406.2 | NP_034536.1 | 3 | 330 | ||
GenBank mRNA | AK080166.1 | - | 3 | 329 | ||
M35525.1 | - | 3 | 285 | |||
M35526.1 | - | 3 | 285 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010406.2 | NP_034536.1 | 3 | 330 | ||
GenBank mRNA | AK080166.1 | - | 3 | 329 | ||
M35525.1 | - | 3 | 285 | |||
M35526.1 | - | 3 | 285 |