siRNA Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010431.2 | NP_034561.2 | 2 | 600 | ||
| GenBank mRNA | AF003695.1 | - | 2 | 434 | ||
| AK017853.1 | - | 2 | 461 | |||
| AK034087.1 | - | 2 | 566 | |||
| AK138944.1 | - | 2 | 467 | |||
| AK150367.1 | - | 2 | 467 | |||
| BC026139.1 | - | 2 | 452 | 4019056 | ||
| BY725437.1 | - | 2 | 600 | |||
| X95002.1 | - | 1 | 114 | |||
| X95580.1 | - | 2 | 266 | |||

Hif1a
Genomic Map