siRNA Details
| Gene Symbol: Hoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010451.2 | NP_034581.1 | 2 | 1534 | ||
| GenBank mRNA | AK157713.1 | - | 2 | 1149 | ||
| M95599.1 | - | 2 | 1534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxa2| Gene Symbol: Hoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010451.2 | NP_034581.1 | 2 | 1534 | ||
| GenBank mRNA | AK157713.1 | - | 2 | 1149 | ||
| M95599.1 | - | 2 | 1534 | |||