siRNA Details
| Gene Symbol: Hoxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010453.5 | NP_034583.1 | 1, 2 | 806 | ||
| GenBank mRNA | AB221550.1 | - | 1, 2 | 569 | ||
| AK144356.1 | - | 1, 2 | 547 | |||
| BC011063.1 | - | 1, 2 | 630 | 3985274 | ||
| M28021.1 | - | 1, 2 | 806 | |||
| M36604.1 | - | 1, 2 | 683 | |||
| X16840.1 | - | 1, 2 | 683 | |||

Hoxa5
Genomic Map