siRNA Details
| Gene Symbol: Hoxc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010466.2 | NP_034596.1 | 2 | 701 | ||
| GenBank mRNA | AB221617.1 | - | 2 | 464 | ||
| AK051173.1 | - | 2 | 157 | |||
| AK079948.1 | - | 2 | 290 | |||
| AK080107.1 | - | 2 | 263 | |||
| AK083688.1 | - | 1 | 699 | |||
| X07439.1 | - | 2 | 701 | |||
| X07646.1 | - | 2 | 929 | |||

Hoxc8
Genomic Map