siRNA Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010475.1 | NP_034605.1 | 1, 2 | 128 | ||
| GenBank mRNA | AK143346.1 | - | 1, 2 | 77 | ||
| BC125657.1 | - | 1, 2 | 127 | 40130172 | ||
| BC125659.1 | - | 1, 2 | 127 | 40130174 | ||
| X89627.1 | - | 1, 2 | 128 | |||

Hsd17b1
Genomic Map