siRNA Details
| Gene Symbol: Hspd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010477.4 | NP_034607.3 | 3 | 447 | ||
| GenBank mRNA | AK088844.1 | - | 3 | 361 | ||
| AK143882.1 | - | 3 | 352 | |||
| AK146831.1 | - | 3 | 399 | |||
| AK219639.1 | - | 2 | 191 | |||
| BC016400.1 | - | 3 | 333 | 3585270 | ||
| BC106112.1 | - | 3 | 348 | |||
| X55023.1 | - | 3 | 309 | |||

Hspd1
Genomic Map