siRNA Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010485.3 | NP_034615.2 | 2 | 364 | ||
| GenBank mRNA | AK034536.1 | - | 2 | 351 | ||
| AK075818.1 | - | 2 | 326 | |||
| AK078333.1 | - | 2 | 680 | |||
| AK080365.1 | - | 2 | 322 | |||
| AK081575.1 | - | 1 | 3855 | |||
| AK089088.1 | - | 1 | 71 | |||
| AK148528.1 | - | 2 | 292 | |||
| AK161667.1 | - | 1 | 81 | |||
| AK180399.1 | - | 1 | 105 | |||
| AK180790.1 | - | 1 | 105 | |||
| AK195786.1 | - | 1 | 66 | |||
| AK197032.1 | - | 1 | 105 | |||
| AK204358.1 | - | 1 | 49 | |||
| U65735.1 | - | 1 | 133 | |||

Elavl1
Genomic Map