siRNA Details
| Gene Symbol: Elavl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010485.3 | NP_034615.2 | 2 | 389 | ||
| GenBank mRNA | AK034536.1 | - | 2 | 376 | ||
| AK075818.1 | - | 2 | 351 | |||
| AK078333.1 | - | 2 | 705 | |||
| AK080365.1 | - | 2 | 347 | |||
| AK081575.1 | - | 1 | 3880 | |||
| AK089088.1 | - | 1 | 96 | |||
| AK148528.1 | - | 2 | 317 | |||
| AK161667.1 | - | 1 | 106 | |||
| AK180399.1 | - | 1 | 130 | |||
| AK180790.1 | - | 1 | 130 | |||
| AK190746.1 | - | 1 | 130 | |||
| AK197032.1 | - | 1 | 130 | |||
| AK198784.1 | - | 1 | 81 | |||
| AK203188.1 | - | 1 | 130 | |||
| U65735.1 | - | 1 | 158 | |||

Elavl1
Genomic Map