siRNA Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB303567.1 | - | 3 | 211 | ||
| AK206264.1 | - | 2 | 80 | |||
| BC002030.1 | - | 4 | 334 | 3487814 | ||
| U26459.1 | - | 4 | 323 | |||
| U26460.1 | - | 3 | 229 | |||
| U26461.1 | - | 3 | 228 | |||
| U34595.1 | - | 2 | 196 | |||

Ica1
Genomic Map