siRNA Details
| Gene Symbol: Ica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252266.1 | NP_001239195.1 | 6 | 756 | ||
| NM_010492.3 | NP_034622.3 | 7 | 1169 | |||
| GenBank mRNA | AB303567.1 | - | 6 | 680 | ||
| AK016989.1 | - | 7 | 872 | |||
| BC002030.1 | - | 7 | 803 | 3487814 | ||
| CB056959.1 | - | 2 | 266 | |||
| DV658174.1 | - | 1 | 79 | |||
| DV659030.1 | - | 5 | 541 | |||
| U26459.1 | - | 7 | 792 | |||
| U26460.1 | - | 6 | 698 | |||
| U26461.1 | - | 6 | 697 | |||
| U34595.1 | - | 5 | 665 | |||

Ica1
Genomic Map