siRNA Details
| Gene Symbol: Ifit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010501.2 | NP_034631.1 | 2 | 199 | ||
| GenBank mRNA | AK077459.1 | - | 2 | 197 | ||
| AK151032.1 | - | 2 | 199 | |||
| AK153226.1 | - | 2 | 200 | |||
| BC003804.1 | - | 2 | 165 | 3487345 | ||
| BC089563.1 | - | 2 | 197 | 30317428 | ||
| CT010266.1 | - | 1 | 128 | |||
| L32974.1 | - | 2 | 180 | |||

Ifit3
Genomic Map