siRNA Details
| Gene Symbol: Igf2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010515.2 | NP_034645.2 | 3, 4 | 576 | ||
| GenBank mRNA | BB865604.1 | - | 3, 4 | 526 | ||
| BC150817.1 | - | 3, 4 | 487 | 9087728 | ||
| BC171973.1 | - | 3, 4 | 487 | 9054259 | ||
| U04710.1 | - | 3, 4 | 532 | |||

Igf2r
Genomic Map