siRNA Details
| Gene Symbol: Ikbkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010546.2 | NP_034676.1 | 21 | 2497 | ||
| GenBank mRNA | AF088910.1 | - | 21 | 2557 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ikbkb| Gene Symbol: Ikbkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010546.2 | NP_034676.1 | 21 | 2497 | ||
| GenBank mRNA | AF088910.1 | - | 21 | 2557 | ||