siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010560.3 | NP_034690.3 | 4 | 459 | ||
| GenBank mRNA | AK170020.1 | - | 5 | 621 | ||
| BC020454.1 | - | 4 | 413 | |||
| BC058679.1 | - | 4 | 420 | 6834623 | ||
| M83336.1 | - | 3 | 264 | |||
| X62646.1 | - | 3 | 264 | |||

Il6st
Genomic Map