siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010560.3 | NP_034690.3 | 6 | 851 | ||
| GenBank mRNA | BC020454.1 | - | 6 | 805 | ||
| BC058679.1 | - | 6 | 812 | 6834623 | ||
| M83336.1 | - | 5 | 656 | |||
| X62646.1 | - | 5 | 656 | |||

Il6st
Genomic Map