siRNA Details
| Gene Symbol: Epb4.1l4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013512.2 | NP_038540.2 | 3 | 914 | ||
| GenBank mRNA | AK140485.1 | - | 2 | 598 | ||
| AK161182.1 | - | 3 | 674 | |||
| BC007166.1 | - | 3 | 638 | 3490061 | ||
| BC011099.1 | - | 2 | 233 | |||
| BC013557.1 | - | 3 | 638 | 3490326 | ||
| BC089158.1 | - | 2 | 505 | |||
| D28818.1 | - | 2 | 188 | |||

Epb4.1l4a
Genomic Map