siRNA Details
| Gene Symbol: Fcer2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253737.1 | NP_001240666.1 | 11 | 1536 | ||
| NM_001253739.1 | NP_001240668.1 | 11 | 1547 | |||
| NM_001253743.1 | NP_001240672.1 | 12 | 1596 | |||
| NM_001253745.1 | NP_001240674.1 | 10 | 1473 | |||
| NM_001253746.1 | NP_001240675.1 | 10 | 1473 | |||
| NM_001253747.1 | NP_001240676.1 | 11 | 1557 | |||
| NM_013517.4 | NP_038545.1 | 12 | 1677 | |||
| GenBank mRNA | M34163.1 | - | 12 | 1675 | ||
| M99371.1 | - | 12 | 1669 | |||

Fcer2a
Genomic Map