siRNA Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005065.5 | NP_005056.3 | 21 | 6901 | ||
| GenBank mRNA | AB020335.1 | - | 21 | 6861 | ||
| CB986742.1 | - | 1 | 132 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEL1L| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005065.5 | NP_005056.3 | 21 | 6901 | ||
| GenBank mRNA | AB020335.1 | - | 21 | 6861 | ||
| CB986742.1 | - | 1 | 132 | |||